Would you like to log in again?
Pipettes, Transfer, Volumetric, Class A, Individually Certified, United Scientific Supplies
Supplier: United Scientific Supplies
PT7100-G PT7100-F PT7100-D PT7100-B PT7100-E PT7100-C PT7100-H PT7100-A
76532-134PK 94.4 USD InStock
76532-134 76532-132 76532-144 76532-138 76532-136 76532-130 76532-142 76532-140
Pipettes, Transfer, Volumetric, Class A, Individually Certified, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
Volumetric pipettes with color-coding.
  • Printed with durable amber markings
  • Color-coded in blue for easy identification of capacity
Made from borosilicate glass in compliance with ASTM E969, Class A standards. Individually serialized and certified; calibrated to deliver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty