Would you like to log in again?
Pipettes, Transfer, Volumetric, Class A, Batch Certified, United Scientific Supplies
Supplier: United Scientific Supplies
PT7101-H PT7101-G PT7101-C PT7101-E PT7101-B PT7101-A PT7101-D PT7101-F
76532-156PK 48.64 USD InStock
76532-156 76532-146 76532-154 76532-158 76532-148 76532-160 76532-152 76532-150
Pipettes, Transfer, Volumetric, Class A, Batch Certified, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
Volumetric pipettes with color-coding.
  • Printed with durable amber markings
  • Color-coded in blue for easy identification of capacity
Made from borosilicate glass in compliance with ASTM E969, Class A standards. Batch certified; calibrated to deliver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty