Would you like to log in again?
Pipettes, Transfer, Volumetric, Class B, United Scientific Supplies
Supplier: United Scientific Supplies
PT7102-D PT7102-A PT7102-F PT7102-C PT7102-H PT7102-G PT7102-B PT7102-E
76532-168PK 27.4 USD InStock
76532-168 76532-176 76532-166 76532-170 76532-174 76532-164 76532-172 76532-162
Pipettes, Transfer, Volumetric, Class B, United Scientific Supplies
Pipettes Pasteur/Transfer Pipettes
Volumetric pipettes with color-coding.
  • Printed with durable amber markings
  • Color-coded in blue for easy identification of capacity
Made from borosilicate glass in compliance with ASTM E969, Class B standards; calibrated to deliver.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty