Would you like to log in again?
Test Tubes with Rim
Supplier: United Scientific Supplies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
TT9800-M TT9800-J TT9800-L TT9800-G TT9800-C TT9800-D TT9800-B TT9800-E TT9800-H TT9800-I TT9800-A TT9800-F TT9800-N TT9800-K
76532-200PK 25.56 USD InStock
76532-200 76532-212 76532-198 76532-210 76532-204 76532-216 76532-202 76532-214 76532-196 470123-578 76532-208 76532-206 470123-580 76532-218
Test Tubes with Rim
Tubes Test Tubes
Pre-cleaned borosilicate glass test tubes with rims. Tubes can be used for hot or cold applications, are reusable, and can be autoclaved.
  • Tubes have rims
  • Autoclavable
Made from borosilicate glass, these general-purpose tubes will withstand daily use at an affordable price. Tubes have a uniform thickness and a strong lip.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty