Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Temperature-Controlled GenoLyte® Compact Tissue Homogenizer and Cell Lyser, HG-250
Supplier: SPEX SamplePrep
1200C
76533-052EA 8094 USD InStock
76533-052
Temperature-Controlled GenoLyte® Compact Tissue Homogenizer and Cell Lyser, HG-250
Homogenizers Benchtop Homogenizers
A powerful, compact, temperature controlled homogenizer.
  • Accommodates most standard 2 ml vials
  • Keep an eye on the grinding process with the clear lid for viewing
  • Cycles can be programmed with rest periods
  • Ideal for applications involving sample prep for DNA, RNA, and protein extractions
The Chilled GenoLyte® is an efficient, compact laboratory homogenizer that uses a circulating chiller to maintain sample temperatures between 0 to 10 °C while effectively disrupting cellular materials by horizontally oscillating 2 ml vials. Common samples include seeds, stems, roots, leaves, and animal tissue.
Operation is simple: The vials are secured in the sample cooling chamber, and the lid is closed. The controls are checked for the proper running time, rate, and the start button is pushed. When the run is complete, the lid is lifted and the vials are removed. The digital control panel allows the user to set the timer in five-second increments; the timer sounds down in one-second intervals.
The vial holder accommodates three vials that are continuously cooled. A circulating chiller is required for continuous temperature control. The GenoLyte is compatible with Polystat® Circulating Baths or most circulating chiller systems.
Ordering information: Purchase chiller and vials separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty