Would you like to log in again?
Cole-Parmer® SM-300 ShatterBox® Verastile Ring and Puck Mills
Supplier: SPEX SamplePrep
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Cole-Parmer®
8521-H 8501-H 8504-H 8506-H 8550-115 8550-230 8505-H
76533-154EA 3349.48 USD InStock
76533-154 76533-156 76533-058 76533-158 76533-060 76533-160 76533-162
Cole-Parmer® SM-300 ShatterBox® Verastile Ring and Puck Mills
Mills
Grind your samples with minimal loss in a soundproof enclosure.
  • Store up to 10 different grinding programs
  • Robust IP 65 rated industrial interface
  • Accommodates various grinding dishes
  • Sound-insulated steel cabinet
  • Electronically controlled, fully reproducible grinding methods with Ethernet capabilities
The Cole-Parmer® SM-300 ShatterBox® Verastile Ring and Puck Mill can crush and grind many types and sizes of samples with minimal sample loss and increased safety. It features an automated locking dish and guided dish positioning with an automatic elevation of the dish for ease of use and greater flexibility. The versatility of sample containers allows for grinding within smaller dishes without the ring as well as the ability to increase sample sizes and capacity with the ring and puck system.
The ShatterBox swings a dish-shaped grinding container with a puck and ring inside in a tight high-speed circle. The sample is quickly crushed between the wall and floor of the container and the moving puck/ring grinding elements. Grinding times range from 2 to 5 minutes, with resultant particle size well below 100 µg/ml, and in some cases 5 µg/ml.
Typical samples include cement, superalloy, granite, glass, sand, slag, ceramics, seeds, grains, pigments, and catalyst supports.
The ShatterBox requires use of a dish with a handle assembly, which is available in hardened steel, tungsten carbide, alumina ceramic, and zirconia.
Ordering information: Supplied without grinding dish, which must be ordered separately.
Delivery information: Supplied with ShatterBox, instruction manual, USB memory stick, 10 mm wrench, Torx keys, and power cable.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty