Would you like to log in again?
Accessories for Matrix Orbital Delta Plus and Matrix Delta Plus Thermoshakers
Supplier: IKA
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0030001244 0030000853 0030000848 0030001158 0030000863 0030000859 0030000829 0030000834 0030001159 0030001253 0030000849 0030001157 0030000821 0030000844 0030000840 0030000857
76533-326EA 1165.93 USD InStock
76533-326 76533-338 76533-316 76533-328 76533-318 76533-340 76533-330 76533-320 76533-342 76533-332 76533-344 76533-322 76533-334 76533-324 76533-336 76533-314
Accessories for Matrix Orbital Delta Plus and Matrix Delta Plus Thermoshakers
Shakers and Mixers Shaking Incubators
The ideal attachment design ensures an optimal temperature transfer and thus guarantees valid results. The thermoshakers Matrix Orbital Delta plus and Matrix Delta plus automatically recognize, which attachment is connected to the device via an interface in the locking mechanism. The attachment can be changed quickly and easily with an ergonomic lever.
  • The heatable ThermoCover prevents condensation on the vial lid and wall. At the same time it enables constant reaction conditions
As part of IKA's Integrated Safety Concept, the insulating plastic coating of the attachment ensures user safety, as high temperatures are not transferred to the frame.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty