Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Puro-i Primary Grade RO Lab Water System, Avidity Science

Supplier: Avidity Sciences
7120-2500-472 7120-2500-492
76533-362EA 8393.19 USD InStock
76533-362 76533-364
Puro-i Primary Grade RO Lab Water System, Avidity Science
Water Purification Systems Deionizers
Provides Type 3 - Reverse osmosis water.

  • Fast, volumetric and variable dispense up to 2 L/min
  • Make-up flow rate of 10 L/h
  • Remote dispensing option for Type 3 water
  • Unique twin pass RO technology
  • Auto RO santization
  • Interactive 7" touch screen
  • Auto reservoir sanitization option
  • Flexible pre-treatment options for variable feed water quality
  • Choice of storage reservoir size - 35, 70, or 105 L

The Puro-i 10 utilizes well established water purification technologies which you can trust to provide you with the qaulity of water you require, on request. Call for water from the dispense tap on the front of the machine to obtain the concise water quality you need; when you need it.

The unique reverse osmosis (RO) cartirdges have a built-in pre-treatment which only requires changing once per year. It means that the water is always in optimum conditions as there is no deterioration of quality from an RO which is several years old.

Application: This model takes potable water to Type 3 at a rate of 10 liters per hour which can be dispensed from a 35 liter, 70 liter, or 105 liter reservoir.

Certifications: Manufactured to ISO 9001:2015 standard. Certified to IEC 61010-1:2010 and EN 61326-21:2013.

Ordering information: Endotoxin and Nuclease-free point of use filter available (see 36337-284).

Delivery information: First round of consumables are included with purchase of unit. Ships within 2 to 3 weeks and installed within 4 to 6 weeks from receiving the PO.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty