Would you like to log in again?
Essentials Slot Samplers, Metal, Antylia Scientific
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Essentials
06286-04 06286-11 06286-07 06286-09 06286-06 06286-12 06286-08 06286-10 06286-03 06286-05
76533-692EA 380.84 USD InStock
76533-692 76533-682 76533-684 76533-674 76533-686 76533-676 76533-688 76533-678 76533-690 76533-680
Essentials Slot Samplers, Metal, Antylia Scientific
Samplers Solid Samplers
Reliably sample various powders, granules, and grains.
  • Suitable for sampling from all layers of the container, providing an accurate representation of contents
  • Choose from stainless steel or aluminum construction
  • Nonsterile
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. These samplers come in different lengths, with different numbers of openings, and are made from different material options for all different sampling methods.
The 316L stainless steel construction is durable and ideal when chemical resistance is required. The lightweight aluminum construction means even the long samplers are easy to operate and manage.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty