Would you like to log in again?
Essentials Sleeve Sample Chamber and Handle, Antylia Scientific
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Essentials
06286-16 06286-15
76533-698EA 313.62 USD InStock
76533-698 76533-700
Essentials Sleeve Sample Chamber and Handle, Antylia Scientific
Samplers Fluid Samplers
Reliably sample large volumes at great depths.
  • Designed to be extremely strong and robust to allow samples to be taken from depths up to 6 meters
  • Constructed of durable 316L stainless steel
  • Non sterile
The sleeve sampler is ideal for sampling large volumes of powder or granules at a depth of up to 6 meters. The heavy duty 316L stainless steel handle provides enhanced strength and durability. Simply push the sleeve sampler into the powder or granules to the required depth and pull up on the handle to open the sleeve and take a sample. Sleeve sampler handle and chamber sold separately.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty