Would you like to log in again?
Essentials Double-Ended Spoons, 304 Stainless Steel, Antylia Scientific
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Essentials
06700-72 06700-70 06700-73 06700-74 06700-71
76533-780EA 14.06 USD InStock
76533-780 76533-782 76533-784 76533-786 76533-778
Essentials Double-Ended Spoons, 304 Stainless Steel, Antylia Scientific
Spoons
Take different sample sizes from the same spoon.
  • 304 stainless steel (SS) construction for excellent chemical compatibility
  • BSE/TSE free
  • Non sterile
Simply flip around the spoon to take different sample sizes. Spoons are constructed of 304 SS with a polished smooth finish—ideal for powders and liquid.
Certifications: Supplier Certificate of Conformity is included with product shipment.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty