Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Essentials Pharma Scoops, 316L Stainless Steel, Antylia Scientific

Supplier: Antylia Scientific
Essentials
07205-12 07205-11 07205-13 07205-20 07205-16 07205-14 07205-17 07205-10 07205-18 07205-19 07205-15
76533-792EA 119.69 USD InStock
76533-792 76533-808 76533-794 76533-796 76533-798 76533-788 76533-800 76533-802 76533-804 76533-790 76533-806
Essentials Pharma Scoops, 316L Stainless Steel, Antylia Scientific
Scoops
High-quality 316L stainless steel (SS) scoops with no dead spaces or crevices.

  • Smooth, mirror polish surface—better than 0.5 µm Ra is ideal for powders and granules
  • Heavy gauge 316L SS offers excellent corrosion resistance and strength
  • BSE/TSE free

These easy-to-clean 316L SS scoops feature hygienic design ideal for the pharmaceutical industry. Fully welded, high-quality scoops have a smooth surface and no crevices where powders and granules can get stuck. Available in 11 sizes for all types of work from the laboratory to production.

Certifications: Supplier Certificate of Conformity is included with product shipment.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty