Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Micro Star 21 and 21R Microcentrifuges
Supplier: VWR
VWR®
75007245 75007244
76533-878EA 6225.95 USD InStock
76533-878
VWR® Micro Star 21 and 21R Microcentrifuges
Centrifuges Microcentrifuges
VWR® Micro Star 21 ventilated and 21R refrigerated microcentrifuge with 24×1.5/2.0 ml rotor with ClickSeal™ biocontainment lid offer exceptional acceleration of up to 21000 xg in as little as 12 seconds.
  • Durable and flexible rotors
  • Outstanding corrosion resistance with lightweight engineered polymer rotors
  • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 ml tubes to mini-preps and spin columns
  • ClickSeal™ Biocontainment solution gives ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with ClickSea™l biocontainment lids
  • Quiet, easy to use design means simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure
  • Sleek, lightweight, space-saving design is durable and easy to clean and maintain
  • Quiet performance promotes a safe, stress-free environment
Refrigerated version offers cooling from room temperature to set temperature in as little as 9 minutes.
Centrifuges have a 2 year warranty. Rotors have a different warranty period.
Certifications: ANSI/UL 61010-1 3.1 Edition
UL 61010-2-020 3rd Edition
UL 61010-2-011 2nd Edition
UL 61010-2-101 3rd Edition
FCC Part 15
ICES-001
EN ISO 14971
ISO 9001
Delivery information: Units supplied with 24×1.5/2.0 ml rotor with ClickSeal biocontainment lid as standard.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty