Would you like to log in again?
Cole-Parmer® Essentials Straight-Sided Jars, Amber Glass, Antylia Scientific
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
34700-53 34700-65 34700-57 34700-66
76533-906CS 53.74 USD InStock
76533-906 76533-908 76533-902 76533-904
Cole-Parmer® Essentials Straight-Sided Jars, Amber Glass, Antylia Scientific
Jars General Use Jars
Don't strain to get samples.
  • Easy access to contents with wide mouth design
  • Store light-sensitive items safely in amber jars
  • Pulp/vinyl lined screw caps protect contents
Manufactured from type III soda-lime glass, these wide mouth jars allow easy access when adding and removing contents. Ideal for general lab use, amber jars are best for light sensitive samples.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty