Would you like to log in again?
Cole-Parmer® Essentials Pre-Cleaned EPA Wide Mouth Jars, Clear Glass, Antylia Scientific
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
99540-42 99540-40 99540-41 99540-44 99540-27 99540-43
76533-970CS 39.07 USD InStock
76533-970 76533-972 76533-974 76533-966 76533-968 76533-948
Cole-Parmer® Essentials Pre-Cleaned EPA Wide Mouth Jars, Clear Glass, Antylia Scientific
Jars General Use Jars
Certified clean for worry-free sampling.
  • Includes certificate of conformance to EPA specifications
  • Leakproof PTFE-lined caps protect contents
  • Identify samples easier with included analysis labels (unattached)
  • Autoclavable
These pre-cleaned jars are processed and certified to meet the requirements of the current US EPA 'Specifications and Guidance for Contaminant-free Sample Containers' document for semi-volatile organics, pesticides, PCBs, metals, cyanide, fluoride, nitrate, and nitrite analyzes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty