Would you like to log in again?
Cole-Parmer® Essentials Heavy-Duty Plastic Bottles, HDPE, Antylia Scientific
Supplier: Antylia Scientific
06025-60 06025-50 06025-70
76534-008PK 120.84 USD InStock
76534-008 76534-012 76534-010
Cole-Parmer® Essentials Heavy-Duty Plastic Bottles, HDPE, Antylia Scientific
Bottles Plastic Bottles
Strong, leakproof storage.
  • Polypropylene closure will not leak
  • Stands up to general storage or shipping
Leakproof and rugged, these high-density polyethylene (HDPE) bottles make excellent storage or shipping containers for liquids or dry chemicals. Bottles are not autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty