Would you like to log in again?
Cole-Parmer® Essentials Economy Narrow-Mouth Plastic Bottles, PPCO, Antylia Scientific
Supplier: Antylia Scientific
06034-38 06034-30 06034-26 06034-34 06034-44
76534-062PK 41.2 USD InStock
76534-062 76534-060 76534-058 76534-056 76534-054
Cole-Parmer® Essentials Economy Narrow-Mouth Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Store chemical samples safely.
  • Leakproof, polypropylene caps
  • Built-in shoulder loop for easier handling
  • Autoclavable
Use these sturdy bottles whenever excellent chemical resistance is desired for most acids, bases, and alcohols. Ideal for storing chemicals and distilled water. Larger bottles feature built-in shoulder loop.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty