Would you like to log in again?
Cole-Parmer® Essentials Economy Wide-Mouth Plastic Bottles, PPCO, Antylia Scientific
Supplier: Antylia Scientific
06034-73 06034-53 06034-71 06034-59 06034-63 06034-51 06034-67 06034-69 06034-55 06034-61 06034-57 06034-65
76534-084PK 46.3 USD InStock
76534-084 76534-074 76534-082 76534-072 76534-080 76534-070 76534-068 76534-066 76534-078 76534-086 76534-064 76534-076
Cole-Parmer® Essentials Economy Wide-Mouth Plastic Bottles, PPCO, Antylia Scientific
Bottles Plastic Bottles
Save money while keeping samples contained.
  • Lightweight design minimizes added weight
  • Leakproof PP closures
  • Autoclavable
These lightweight bottles are suitable for most laboratory applications. Ideal for light-duty, general-purpose applications where heavier bottles are not required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty