Would you like to log in again?
Cole-Parmer® Cryogenic Vials, External Thread, Sterile, Antylia Scientific
Supplier: Antylia Scientific
78903-21 78903-20
76534-406CS 251.62 USD InStock
76534-406 76534-404
Cole-Parmer® Cryogenic Vials, External Thread, Sterile, Antylia Scientific
Vials
Stores samples as low as –196 °C.
  • DNase-, RNase- and pyrogen-free
  • Radiation sterilized
  • HDPE cap
These polypropylene cryogenic vials feature a self-standing star base designed for easy one-handed manipulation when used with a support rack.
Vials include an internal U-shaped bottom, large white frosted labeling area, silk screen graduations and a leak-proof HDPE cap.
Ideal for used in vapor phase of liquid nitrogen but are not recommended for use in the liquid phase of liquid nitrogen.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty