Would you like to log in again?
Cole-Parmer® Essentials Disposable Inoculation Needles, Antylia Scientific
Supplier: Antylia Scientific
06231-06 06231-01 06231-09
76534-526CS 80.04 USD InStock
76534-526 76534-524 76534-522
Cole-Parmer® Essentials Disposable Inoculation Needles, Antylia Scientific
Spreaders Inoculating Loops and Needles
Save time and eliminate cross-contamination.
  • Constructed of durable polystyrene
Smooth and flexible needles are constructed of high-impact polystyrene. Straight needles are ideal for picking single colonies. Color coding identifies inoculation volume easily.
Certifications: Each case is supplied with a quality certificate confirming sterility and accuracy.
Packaging: Packed sterile in tamperproof resealable bags.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty