Would you like to log in again?
Argos Technologies® Reusable Inoculating Loop and Needle Holders, Antylia Scientific
Supplier: Antylia Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
04396-40 04396-41
76534-528EA 50.66 USD InStock
76534-528 76534-530
Argos Technologies® Reusable Inoculating Loop and Needle Holders, Antylia Scientific
Spreaders Inoculating Loops and Needles
Save more with these reusable loop and needle handles.
  • Handles (6 or 8") are sold separately for some loops and needles
Reusable Inoculating Loops with handles are used to select, inoculate or transfer microbiological samples. Handles are insulated to protect the hand during use.
Loops and Handles cool rapidly after sterilization.
Ordering information: Loops and without handles are sold separately as models 04396-35 through -39. Needle without handle is available as model 04396-49.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty