Would you like to log in again?
Argos Technologies® Reusable Inoculating Streakers, Antylia Scientific
Supplier: Antylia Scientific
04396-43 04396-44 04396-42 04396-46 04396-45
76534-548PK 330.66 USD InStock
76534-548 76534-546 76534-544 76534-542 76534-550
Argos Technologies® Reusable Inoculating Streakers, Antylia Scientific
Spreaders Inoculating Loops and Needles
Nichrome wire composition cools quickly and is less expensive than platinum.
  • Select, inoculate, or transfer microbiological samples
Loops are made of high quality twisted nichrome wire and attached to a light-weight aluminum handle. Handles are insulated to protect the hand during use. Loops have been designed to slide along agar surfaces without cutting or tearing.
Loops and Handles cool rapidly after sterilization and are offered calibrated (1 and 10 µl) or non-calibrated. Non-calibrated Loops and Handles are available as small, medium or large and are color coded for easy identification.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty