Would you like to log in again?
Thermo Scientific™ Conical Centrifuge Tubes, Thermo Scientific
Supplier: Thermo Scientific
332270 332270 332265 332265
76534-586CS 316.7 USD InStock
CA76533-866 76534-586 CA76533-868 76534-584
Thermo Scientific™ Conical Centrifuge Tubes, Thermo Scientific
Tubes Centrifuge Tubes
The Thermo Scientific™ Conical Centrifuge Tubes are sterile and ideal for applications requiring larger sample volumes.
  • Non-pyrogenic, non-cytotoxic
  • DNase/RNase, BSE/TSE, human DNA free
Manufactured with high quality virgin resins.
Certifications: Sterilized by gamma irradiation per ISO 11137-2.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty