Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CellXpert® C170i, CO₂ Incubators

Supplier: Eppendorf
CellXpert®
6731010045 6731010515 2231000760 6731011015 2231000762 6731010355 2231000664 6731010845 6731010015 6731010345 6731010215 2231000760 6731011055 6731011815 6731011045 6731010045 2231000763 6731010515 6731010215 6731010225 2231000760 2231000763 2231001051 6731010055 6731011525 6731010125 6731010115 6731010055 2231001051 2231000763 6731011515 2231000762 2231000761 2231000762 6731010025 6731012085 6731010525 6731011355 6731010315 2231000851 2231000850 6731012045 6731011825 6731010525 6731011025 6731010855 2231000761 6731010015 6731012045 6731010325 6731010855 6731010355 6731010845 6731010315 6731011025 6731011515 6731010345 6731010125 6731010115 6731010225 6731011525 6731011345 6731011355 6731010325 6731011055 6731010025 6731011345 6731011045
76534-598EA 11149.88 USD InStock
CA76468-862 76534-598 CA76308-814 76534-596 CA76308-816 CA77321-636 76534-594 CA77321-638 77321-634 77321-636 CA77321-640 76534-622 CA76534-598 CA77321-642 MPCA76534-608 MPCA76534-606 CA77321-648 CA76534-604 77321-648 CA76534-602 CA77321-650 77321-642 CA76534-600 MPCA76308-816 MPCA76308-815 77321-640 CA76534-608 MPCA76460-805 CA76534-606 MPCA76460-809 76308-820 MP76468-918 76460-804 76534-610 76468-918 76534-606 CA76534-616 76534-604 CA76534-614 MP-76308-814 CA76534-612 76534-602 CA76534-610 76534-600 MP76468-920 MPCA76460-811 77321-650 CA76534-618 76534-608 MP-76468-920 CA76308-820 76468-920 MP76308-816 76308-816 CA76534-596 76308-818 CA76534-594 76308-814 76460-810 CA77321-634 76534-616 76460-808 76534-614 76460-806 CA76534-620 76534-612 CA76468-860
CellXpert® C170i, CO₂ Incubators
Incubators CO₂ Incubators
This CO₂ incubator makes monitoring and documentation easy, and delivers optimized growth conditions, even for sensitive cells.

  • Easy cleaning and disinfection: Seamless chamber, easy disassembling in 40 seconds, removable water tray
  • Certified, effective contamination protection: 180 °C High temperature disinfection (HTD) with protocol, no internal fan-associated HEPA filters necessary
  • Uniform temperature: Innovative heating concept with four temperature sensors at various locations verified at 27 locations inside the incubator (German DIN 12880:2007-05)
  • Fast recovery after door opening: Temperature and CO₂ in less than 5 min without regulation overshoot
  • Enhanced vibration and turbulence protection: No fan inside
  • Save money: No internal expendable parts (e.g. fan-associated HEPA filters), significantly reduced gas consumption, up to 25% more usable space by fanless design and innovative insulation concept
  • Stay flexible for the future: Upgrade your device in the laboratory later with various options
  • Full control and easy documentation: Advanced user interface, 2 front USB-ports, customized tasks and alarms, VisioNize® user management and connected lab
  • GMP-/Cleanroom-related certificates and features: ISO class 4/Grade A cleanroom certified (ISO 14644-1), cytotoxicity test of materials: passed (ISO 10993-5:2009), production certificate, pdf event and parameter logging, HTD-effectiveness certificate and more

Easy to handle the new magnetic door locking concept for outer and inner doors. Enhanced vibration and turbulence protection, fan-less design. Effective contamination protection with 180 °C high temperature disinfection (HTD) with protocol, no internal HEPA-filters.

Features customized tasks and alarms to set reminders.

Ordering information: For available accessories, contact your Avantor representative.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty