Would you like to log in again?
Histoware™ Prefilled Specimen Containers, Caplugs
Supplier: Caplugs
308-1109-G8K 308-1101-G8T 308-1107-G8K
76535-038CT 81.58 USD InStock
76535-038 76535-034 76535-036
Histoware™ Prefilled Specimen Containers, Caplugs
Sample Containers General Purpose Containers
Histoware™ line features multiple capacity screw-cap vials prefilled with 10% buffered formalin.
  • Histoware vials nearly transparent and virtually unbreakable
Each size of prefilled vials contains half the nominal capacity of preservative.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty