Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® LED Digital Rotary Evaporators

Supplier: Avantor
VWR®
6030320220 6030320221 6030320222 6030320224 6030320223
76537-058EA 1596.97 USD InStock
76537-058 76533-234 76533-258 76533-260 76533-262
VWR® LED Digital Rotary Evaporators
Evaporators Rotary Evaporators
This is a basic chemical laboratory instrument suitable for evaporation, distillation and separation of chemicals. It can be used as a multifunctional system in combination with a vacuum pump, vacuum controller and refrigerated circulator to provide excellent distillation solutions in many laboratory applications.

  • 5 L heating bath with a wide temperature range (ambient to 180 °C), independent temperature control allows it to be used separately. Water/oil heating mode can only be changed via a switch
  • Manual lift combined with auxiliary lift for precise positioning of the glassware
  • PID control ensures high temperature accuracy at ±1 °C (water)
  • Overheating protection temperature at 220 °C
  • Boil-dry protection, unit automatically powers off if heating without water/oil in the heating bath
  • Speed range from 20 to 200 rpm, timing interval operation in clockwise and anticlockwise directions for drying process
  • Glassware with explosion proof film is available

Evaporators have a patented condenser (cooling surface 1200/1700 cm² for selection), double-helix condensing tube and central circular arc design to speed up the flow rate of liquid with excellent cooling effect. The ejection mechanism ensures easy exchange of evaporating flasks. LED rotary evaporators have an adjustable immersion angle and a PTFE double-spring sealing ring which provides excellent sealing performance. The remote function provides PC control and data transmission, these models are compatible with the entire range of VWR LCD evaporator glassware.

Certifications: Protection class DIN EN 60529 IP 20, IEC 60529

Ordering information: A complete pre-configured system which consists of a main unit, heating bath and set of glassware can be ordered, or alternatively each component can be ordered separately. Accessories are not included and must be ordered separately.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty