Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BRAND® Microcentrifuge Tubes with Attached Lids, 1.5 ml, BrandTech

Supplier: BrandTech

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:


Brand®
780421 MTS15 780425 780420 780422 780424 780423
76537-080PK 38.56 USD InStock
76537-080 76537-082 76537-072 76537-084 76537-074 76537-076 76537-078
BRAND® Microcentrifuge Tubes with Attached Lids, 1.5 ml, BrandTech
Tubes Microcentrifuge Tubes
These 1.5 ml tubes fit most common microcentrifuges.

  • Withstands 20000 RCF
  • Available in clear and five colors
  • Non sterile or sterile

BRAND® microcentrifuge tubes fit most common rotors. These 1.5 ml tubes may be spun up to 20000 RCF (g) at 20 °C for 20 minutes. Manufactured from virgin polypropylene, these highly translucent tubes are free from flash, air bubbles and streaks. Flat caps form a tight, leak-free seal that is easy to open, even when wearing gloves.

Ordering information: Sold in bulk bags.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty