Would you like to log in again?
Linearity FD iPTH for Abbott Systems, AUDIT Microcontrols
Supplier: AUDIT Microcontrols
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
K957M-5
76537-088EA 359.48 USD InStock
76537-088
Linearity FD iPTH for Abbott Systems, AUDIT Microcontrols
Standards Color Reference and Spectroscopy Standards
Linearity FD iPTH for Abbott Systems is a five level linearity set intended to simulate human patient serum samples for purpose of determining linearity, calibration verification, and verification of reportable range for iPTH.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty