Would you like to log in again?
Brady® Standard Key Retaining Lockout Nylon Padlocks with Steel Shackle, Brady
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SDPL-BLU-38ST-KD6 SDPL-RED-38ST-KA6 SDPL-BLU-38ST-KA6 SDPL-BLU-38ST-KD SDPL-RED-38ST-KD6 SDPL-ORG-38ST-KA6 SDPL-RED-38ST-KA24 SDPL-RED-38ST-KD24 SDPL-YLW-38ST-KD SDPL-RED-38ST-KA12 SDPL-GRN-38ST-KD SDPL-RED-38ST-KD12 SDPL-GRN-38ST-KA6 SDPL-GRN-38ST-KD6 SDPL-YLW-38ST-KD6 SDPL-ORG-38ST-KD6 SDPL-ORG-38ST-KD SDPL-RED-38ST-KD SDPL-YLW-38ST-KA6
76537-202PK 120.3 USD InStock
76537-202 76537-204 76537-206 76537-190 76537-180 76537-170 76537-192 76537-182 76537-194 76537-172 76537-184 76537-174 76537-196 76537-186 76537-198 76537-176 76537-188 76537-178 76537-200
Brady® Standard Key Retaining Lockout Nylon Padlocks with Steel Shackle, Brady
Security Locks
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient, and cost-effective Safety Lockout Padlocks from Brady.
  • Standard padlock body type
  • Service temperature range: −15 to +250 °F
Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program. Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
Nylon padlocks with steel shackles are lightweight, durable, and ideal for general lockout applications. Locks that are Keyed Different (KD) are ideal for simple lockout applications and a manageable number of energy isolation points. Record and retain engraved key codes to streamline replacement of lost keys. Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
Certifications: ANSI Z244.1-2003; OSHA 29 CFR 1910.147.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty