Would you like to log in again?
Brady® Standard Key Retaining Lockout Nylon Padlocks with Plastic Shackle, Brady
Supplier: Brady
SDPL-ORG-38PL-KA6 SDPL-RED-38PL-KD SDPL-BLU-38PL-KA6 SDPL-GRN-38PL-KA6 SDPL-YLW-38PL-KD SDPL-YLW-38PL-KD6 SDPL-BLU-38PL-KD6 SDPL-ORG-38PL-KD SDPL-RED-38PL-KD12 SDPL-RED-38PL-KA6 SDPL-RED-38PL-KD6 SDPL-RED-38PL-KA24 SDPL-GRN-38PL-KD SDPL-RED-38PL-KA12 SDPL-GRN-38PL-KD6 SDPL-RED-38PL-KD24 SDPL-BLU-38PL-KD SDPL-YLW-38PL-KA6 SDPL-ORG-38PL-KD6
76537-234PK 62.58 USD InStock
76537-234 76537-212 76537-224 76537-236 76537-214 76537-226 76537-238 76537-216 76537-228 76537-218 76537-208 76537-240 76537-230 76537-220 76537-242 76537-232 76537-210 76537-222 76537-244
Brady® Standard Key Retaining Lockout Nylon Padlocks with Plastic Shackle, Brady
Security Locks
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient, and cost-effective Safety Lockout Padlocks from Brady.
  • Standard padlock body type
  • Service temperature range: −15 to +250 °F
Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program. Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
Nylon padlocks with plastic shackles are lightweight, non-conductive, non-sparking, and ideal for electrical or general lockout applications. Locks that are Keyed Different (KD) are ideal for simple lockout applications and a manageable number of energy isolation points. Record and retain engraved key codes to streamline replacement of lost keys. Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
Certifications: ANSI Z244.1-2003; OSHA 29 CFR 1910.147.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty