Would you like to log in again?
Brady® Standard Key Retaining Lockout Aluminum Padlocks with Steel Shackle, Brady
Supplier: Brady
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SDAL-RED-38ST-KD12 SDAL-GRN-38ST-KD SDAL-RED-38ST-KA6 SDAL-ORG-38ST-KD SDAL-ORG-38ST-KD6 SDAL-YLW-38ST-KD6 SDAL-RED-38ST-KD SDAL-YLW-38ST-KD SDAL-ORG-38ST-KA6 SDAL-BLU-38ST-KD SDAL-BLU-38ST-KD6 SDAL-RED-38ST-KA24 SDAL-RED-38ST-KD24 SDAL-BLU-38ST-KA6 SDAL-RED-38ST-KD6 SDAL-RED-38ST-KA12 SDAL-GRN-38ST-KD6 SDAL-YLW-38ST-KA6 SDAL-GRN-38ST-KA6
76537-246EA 14.42 USD InStock
76537-246 76537-268 76537-256 76537-278 76537-248 76537-258 76537-280 76537-260 76537-282 76537-270 76537-262 76537-250 76537-272 76537-264 76537-252 76537-274 76537-266 76537-276 76537-254
Brady® Standard Key Retaining Lockout Aluminum Padlocks with Steel Shackle, Brady
Security Locks
Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient, and cost-effective Safety Lockout Padlocks from Brady.
  • Standard padlock body type
  • Service temperature range: −15 to +250 °F
Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program. Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
Aluminum padlocks with steel shackles are durable, corrosion-resistant, and ideal for general lockout applications. Locks that are Keyed Different (KD) are ideal for simple lockout applications and a manageable number of energy isolation points. Record and retain engraved key codes to streamline replacement of lost keys. Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.
Certifications: ANSI Z244.1-2003; OSHA 29 CFR 1910.147.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty