Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Muffle Furnaces up to 1200 °C, Models L and LT, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nabertherm
L-014K27N1-240 L-094H2ON2-208 L-154K2LN2-208 L-094K2LN2-240 L-054H2LN2-120 L-244H2LN2-480 L-154K2LN2-240 L-404H2ON2-480 L-054H2ON2-120 L-034K2ON2-208 L-034H2ON2-240 L-034K2LN2-240 L-244K2ON2-480 L-034H2LN2-240 L-054K2ON2-208 L-094H2ON2-240 L-154K2ON2-240 L-154H2LN2-240 L-034H2ON2-120 L-404H2LN2-480 L-154H2ON2-208 L-094K2ON2-240 L-054K2ON2-240 L-054K2LN2-208 L-094H2LN2-208 L-154K2ON2-208 L-034H2ON2-208 L-154H2ON2-240 L-054K2LN2-120 L-034K2LN2-208 L-014K27N1-120 L-054K2ON2-120 L-054K2LN2-240 L-094K2LN2-208 L-054H2LN2-240 L-034H2LN2-120 L-034K2ON2-240 L-054H2LN2-208 L-014K27N1-208 L-034K2ON2-120 L-244K2LN2-480 L-054H2ON2-240 L-054H2ON2-208 L-244H2ON2-480 L-094H2LN2-240 L-094K2ON2-208 L-404K2ON2-480 L-404K2LN2-480 L-034K2LN2-120 L-154H2LN2-208 L-034H2LN2-208
76537-686EA 7852.34 USD InStock
76537-686 76537-642 76537-644 76537-688 76537-720 76537-646 76537-722 76537-648 76537-724 76537-726 76537-728 76537-690 76537-692 76537-650 76537-694 76537-730 76537-696 76537-652 76537-732 76537-698 76537-654 76537-734 76537-656 76537-658 76537-660 76537-662 76537-664 76537-666 76537-668 76537-700 76537-702 76537-704 76537-706 76537-708 76537-670 76537-672 76537-674 76537-710 76537-676 76537-712 76537-678 76537-634 76537-714 76537-636 76537-716 76537-638 76537-718 76537-680 76537-682 76537-684 76537-640
Muffle Furnaces up to 1200 °C, Models L and LT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These muffle furnaces have been proven for daily laboratory use. These models stand out for their workmanship, advanced and attractive design and high level of reliability. The muffle furnaces come equipped with either a fold down or lift up door at no extra charge.

  • Tmax 1200 °C
  • Heating from two sides by ceramic heating plates (heating from three sides for muffle furnaces L 24/11 - LT 40/12)
  • Thermocouple type S; manual or automatic gas supply system
  • Ceramic heating plates with integral heating element which is safeguarded and easy to replace
  • Optional fold down door (L models) which can be used as work platform or lift up door (LT models) with hot surface facing away from the operator
  • Adjustable air inlet integrated in door
  • Exhaust air outlet in rear wall of furnace
  • Controller B510 and C550

Over-temperature limiter with adjustable cutout temperature as temperature limiter to protect the oven and load. Protective gas connection to purge with non-flammable protective or reaction gases (not available in combination with chimney, chimney with fan or catalytic converter) not gas tight. Port for thermocouple in the rear wall or in the furnace door. Charging rack with closed or perforated trays for loading the furnace in two levels including holder for inserting/removing the trays up to a max. temperature of 800 °C.

Accessories information: Chimney, chimney with fan or catalytic converter (not for L 1 and L 15).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty