Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Muffle Furnaces up to 1100 °C, Models L and LT, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nabertherm
L-094K1LN1-240 L-054K1LN1-240 L-034H1LN1-120 L-154H1LN1-240 L-154K1LN1-240 L-404K1LN1-208 L-244K1LN1-480 L-054H1LN1-240 L-404H1LN1-208 L-244H1LN1-480 L-054K1LN1-208 L-154K1LN1-208 L-034H1LN1-208 L-404K1LN1-480 L-054H1LN1-208 L-094K1LN1-208 L-034H1LN1-240 L-094H1LN1-208 L-034K1LN1-208 L-154H1LN1-208 L-034K1LN1-120 L-034K1LN1-240 L-404H1LN1-480 L-054K1LN1-120 L-404K1LN1-208 L-054H1LN1-120 L-094H1LN1-240 L-404H1LN1-208
76537-740EA 4844.74 USD InStock
76537-740 76537-762 CA76639-010 76537-764 76537-742 76537-766 76537-744 CA76639-014 76537-768 76537-746 76537-748 76537-770 76639-010 76537-772 76537-750 76537-774 76537-752 76639-014 76537-776 76537-754 76537-778 76537-756 76537-758 76537-736 76537-738 76537-780 76537-782 76537-760
Muffle Furnaces up to 1100 °C, Models L and LT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These muffle furnaces have been proven for daily laboratory use. These models stand out for their workmanship, advanced and attractive design and high level of reliability. The muffle furnaces come equipped with either a fold down or lift up door at no extra charge.

  • Tmax 1100 °C
  • Heating from two sides by ceramic heating plates (heating from three sides for muffle furnaces L 24/11 - LT 40/12)
  • Thermocouple type S; manual or automatic gas supply system
  • Ceramic heating plates with integral heating element which is safeguarded and easy to replace
  • Optional fold down door (L models) which can be used as work platform or lift up door (LT models) with hot surface facing away from the operator
  • Adjustable air inlet integrated in door
  • Exhaust air outlet in rear wall of furnace
  • Controller B510 and C550

Over-temperature limiter with adjustable cutout temperature as temperature limiter to protect the oven and load. Protective gas connection to purge with non-flammable protective or reaction gases (not available in combination with chimney, chimney with fan or catalytic converter) not gas tight. Port for thermocouple in the rear wall or in the furnace door. Charging rack with closed or perforated trays for loading the furnace in two levels including holder for inserting/removing the trays up to a max. temperature of 800 °C.

Accessories information: Chimney, chimney with fan or catalytic converter (not for L 1 and L 15).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty