Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Muffle Furnaces up to 1400 °C, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nabertherm
L-154V4ON11-240 L-154V4ON11-208 L-154U4ON11-240 L-094U4ON8-240 L-054V4ON10-240 L-094U4ON8-208 L-054V4LN10-240 L-054U4ON10-208 L-154V4LN11-208 L-054V4ON10-208 L-094V4LN8-208 L-054U4LN10-208 L-054V4LN10-208 L-154V4LN11-240 L-154U4ON11-208 L-094V4LN8-240 L-094U4LN8-240 L-054U4ON10-240 L-154U4LN11-240 L-154U4LN11-208 L-054U4LN10-240 L-094U4LN8-208 L-094V4ON8-208 L-094V4ON8-240
76537-796EA 8806.92 USD InStock
76537-796 76537-784 76537-798 76537-820 76537-830 76537-786 76537-822 76537-800 76537-788 76537-810 76537-824 76537-802 76537-812 76537-804 76537-826 76537-814 76537-828 76537-806 76537-816 76537-808 76537-818 76537-790 76537-792 76537-794
Muffle Furnaces up to 1400 °C, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These Muffle furnaces stand out for their excellent workmanship, advanced and attractive design, and high level of reliability. Heating elements on support tubes radiating freely into the furnace chamber provide particularly short heating times and a maximum temperature of 1400 °C. These models are a good alternative to the familiar L(T) ../11 series when higher application temperatures are needed.

  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Dual shell housing for low external temperatures and high stability
  • Solid state relays provide for low-noise operation
  • Defined application within the constraints of the operating instructions
  • NTLog basic for Nabertherm controller: recording of process data with USB-flash drive

The furnaces have heating from two sides, the heating elements on support tubes ensure free heat radiation and a long service life. All models feature an adjustable air inlet which is integrated in the door and an exhaust air outlet in the rear wall of the furnace.

Accessories information: Optional accessories include a chimney, chimney with fan or catalytic converter, over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load, protective gas connection to purge with non-flammable protective or reaction gases (not available in combination with chimney, chimney with fan or catalytic converter), manual or automatic gas supply system, process control and documentation via VCD software package for monitoring, documentation and control.

Ordering information: These muffle furnaces with temperate up to 1400 °C can be ordered in a variety of sizes and voltage options, with fold down door (L models) or lift up door (LT models). All models are rated up to 1400 °C maximum temperature. Control options include the B510 controller (5 programs with 4 segments each) or C550 controller (10 programs with 20 segments each).

Caution: *Temperature uniformity: ± 5 K with closed air inlet in the empty workspace according to DIN 17052-1 at working temperatures > 800 °C when connected at 220-240 V

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty