Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Muffle Furnaces, Models L and LT, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nabertherm
L-094H1ON1-208 L-034K1ON1-240 L-154H1ON1-240 L-404K1ON1-480 L-054K1ON1-240 L-034H1ON1-120 L-154K1ON1-240 L-404H1ON1-208 L-054K1ON1-120 L-094K1ON1-208 L-094H1ON1-240 L-404H1ON1-208 L-094K1ON1-240 L-404K1ON1-208 L-034K1ON1-120 L-054H1ON1-208 L-054H1ON1-240 L-054K1ON1-208 L-154K1ON1-208 L-244H1ON1-480 L-244K1ON1-480 L-034K1ON1-208 L-404K1ON1-208 L-404H1ON1-480 L-034H1ON1-208 L-054H1ON1-120 L-154H1ON1-208
76537-884EA 6963.42 USD InStock
CA76639-012 76537-884 76537-886 76537-888 CA76639-016 76537-868 76537-900 76537-902 76537-904 76537-906 76537-908 76537-890 76537-892 76537-870 76537-894 76639-012 76537-896 76537-874 76537-910 76537-898 76537-876 76639-016 76537-912 76537-878 76537-914 76537-880 76537-882
Muffle Furnaces, Models L and LT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The muffle furnaces L 1/12 - LT 40/12 are the right choice for daily laboratory use.

  • Tmax 1100 °C or 1200 °C
  • Heating from two sides by ceramic heating plates (heating from three sides for the 24/11 - LT 40/12 muffle furnaces)
  • Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
  • Insulation made of non-classified fiber material
  • Housing made of sheets of textured stainless steel
  • Dual shell housing for low external temperatures and high stability
  • Optional fold down door (L models) which can be used as work platform or lift up door (LT models) with hot surface facing away from the operator
  • Adjustable air inlet integrated in door (see illustration)
  • Exhaust air outlet in rear wall of furnace
  • Solid state relays provide for low-noise operation

These models stand out for their excellent workmanship, advanced and attractive design and high level of reliability. The muffle furnaces come equipped with either a flap door or lift door at no extra charge.

Accessories information: Optional accessories include chimney, chimney with fan or catalytic converter; over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; protective gas connection to purge with non-flammable protective or reaction gases; and a manual gas supply system. Contact your Avantor representative for more information.

Ordering information: These muffle furnaces can be ordered in a variety of sizes and voltage options, with fold down door (L) or lift up door (LT). Maximum temperature options of 1100 or 1200 °C. Control options include the B510 controller (5 programs with 4 segments each) or C550 controller (10 programs with 20 segments each). The L 1/12 model comes with the basic 3216 or R7 controller with one set temperature.

Caution: * Temperature uniformity of ± 5K in the empty work space for temperatures >800 °C.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty