Would you like to log in again?
High-Temperature Ovens, Forced Convection Chamber Furnaces, Models N and NA, Nabertherm
Supplier: Nabertherm
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nabertherm
NA--2542M2-480 NA--0161L1-208 NA--1281M2-480 NA--0342K1-480 NA--5042K2-480 NA--1262M2-480 NA--2562M2-480 NA--2581M2-480 NA--2562K2-480 NA--0662K2-480 NA--5062K2-480 NA--0161O1-208 NA--2542K2-480 NAT-0382K1-480 NA--5081M2-480 NA--5042M2-480 NA--0681K2-480 NA--1281K2-480 NA--0161O1-240 NA--2581K2-480 NA--0681M2-480 NA--0342M1-480 NA--0362K2-480 NA--1262K2-480 NA--0161L1-480 NA--0642K2-480 NA--0362M2-480 NA--0642M2-480 NA--1242M2-480 NA--5062M2-480 NAT-0382M1-480 NA--1242K2-480 NA--0662M2-480
76537-960EA 17127.93 USD InStock
76537-960 76537-940 76537-962 76537-920 76537-942 76537-964 76537-922 76537-966 76537-944 76537-968 76537-924 76537-946 76537-948 76537-926 76537-928 76537-970 76537-972 76537-950 76537-952 76537-930 76537-974 76537-932 76537-954 76537-976 76537-978 76537-934 76537-956 76537-958 76537-936 76537-916 76537-938 76537-918 76537-980
High-Temperature Ovens, Forced Convection Chamber Furnaces, Models N and NA, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These chamber furnaces with air circulation are characterized by their extremely high temperature uniformity.
  • Horizontal air circulation, swing door hinged on the right
  • Temperature uniformity up to ±4 °C according to DIN 17052-1 (model NA 15/65 up to ±5 °C)
  • Optimum air flow and temperature uniformity through high circulation rates
  • One frame sheet and rails for two additional trays included in the scope of delivery (NA 15/65 without frame sheet)
  • Stainless steel air-baffles in the furnace for optimum air circulation
  • Base frame included in the delivery, NA 15/65 designed as table-top model
  • Air inlet and exhaust air flaps as additional equipment for using as drying oven
Due to the modular concept, the forced convection furnaces can be adjusted to the process requirements by adding suitable equipment.
Hence, they are especially suitable for processes such as cooling, crystalizing, preheating, curing, but also for numerous processes in tool making.
Accessories information: Optional accessories include: Optimization of the temperature uniformity up to ±3 °C according to DIN 17052-1, air inlet and exhaust air flaps when used for drying; controlled cooling with fan, manual lift door (up to model N(A) 120/.. (HA)), pneumatic lift door, air circulation with speed control, recommendable for processes with light or sensitive charge; additional frame sheet; roller conveyor in furnace chamber for heavy charges, annealing boxes, feed and charging aids, inlets, measuring frames and thermocouples for TUS measurements charge or comparative measurements, and charge control. Contact your VWR representative for more information.
Ordering information: These high-temperature ovens with hot air convection can be ordered in a variety of sizes and maximum temperature options. Models are rated at 450 °C, 650 °C, or 850 °C. Control options include the B-series controller (5 programs with 4 segments each), C-series controller (10 programs with 20 segments each), or the P-series controller (50 programs with 40 segments each).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty