Would you like to log in again?
Chamber Furnaces with Brick Insulation or Fiber Insulation, Nabertherm
Supplier: Nabertherm
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nabertherm
LH0242NMNE-480 LH2142NMNE-480 LH2122NKNE-480 LH1232NMNE-480 LH0642NKNE-480 LF0642NKNE-480 LF0242NKNE-480 LH2122NMNE-480 LF0642NMNE-480 LH0632NMNE-480 LH0332NKNE-480 LH0332NMNE-480 LH0632NKNE-480 LF1242NKNE-480 LH0232NMNE-480 LH0642NMNE-480 LF0232NKNE-480 LF0342NMNE-480 LH2132NKNE-480 LF1232NKNE-480 LH0622NMNE-480 LH0222NMNE-480 LF0342NKNE-480 LH0322NMNE-480 LH1242NKNE-480 LF1242NMNE-480 LF0242NMNE-480 LH1232NKNE-480 LH1222NMNE-480 LH2142NKNE-480 LH0242NKNE-480 LH0222NKNE-480 LH0232NKNE-480 LF1232NMNE-480 LH0322NKNE-480 LF0332NMNE-480 LH0342NKNE-480 LF0632NMNE-480 LH1242NMNE-480 LF0632NKNE-480 LF0232NMNE-480 LH2132NMNE-480 LF0332NKNE-480 LH0342NMNE-480
76538-108EA 19692.29 USD InStock
76538-108 76538-128 76538-106 76538-104 76538-126 76538-148 76538-146 76538-102 76538-124 76538-068 76538-078 76538-132 76538-110 76538-076 76538-098 76538-130 76538-074 76538-096 76538-094 76538-072 76538-092 76538-070 76538-118 76538-138 76538-116 76538-114 76538-136 76538-058 76538-134 76538-112 76538-122 76538-144 76538-100 76538-066 76538-120 76538-142 76538-064 76538-086 76538-140 76538-084 76538-062 76538-060 76538-082 76538-080
Chamber Furnaces with Brick Insulation or Fiber Insulation, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The LH 15/12 - LF 120/14 chamber furnaces have been trusted for many years as professional chamber furnaces for the laboratory.
  • Tmax 1200, 1300, or 1400 °C
  • Dual shell housing with rear ventilation, provides for low shell temperatures
  • Five-sided heating for very good temperature uniformity
  • Heating elements on support tubes ensure free heat radiation and a long service life
  • Controller mounted on furnace door and removable for comfortable operation
  • Protection of bottom heating and flat stacking surface provided by embedded SiC plate in the floor
  • LH models: multi-layered, fiber-free insulation of light refractory bricks and special backup insulation
  • LF models: high-quality non-classified fiber insulation with corner bricks for shorter heating and cooling times
  • Door with brick-on-brick seal, hand fitted
  • Short heating times due to high installed power
  • Self-supporting arch for high stability and greatest possible protection against dust
  • Quick lock on door
  • Motor driven exhaust air flap
  • Freely adjustable air slide intake in furnace floor
  • Base included
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
These furnaces are available with either a robust insulation of light refractory bricks (LH models) or with a combination insulation of refractory bricks in the corners and low heat storage, quickly cooling fiber material (LF models). With a wide variety of optional equipment, these chamber furnaces can be optimally adapted to your processes.
Accessories information: Optional accessories include parallel swinging door, pivots away from operator, for opening when hot; Lift door with electro-mechanic linear drive; Separate wall-mounting or floor standing cabinet for switchgear; Cooling fan for shorter cycle times; Protective gas connection to purge with non-flammable protective or reaction gases; Manual or automatic gas supply system; Scale to measure weight reduction during annealing; Process control and documentation via VCD software package or Nabertherm Control Center (NCC) for monitoring, documentation and control. Contact your VWR representative for more information.
Ordering information: These chamber furnaces with brick (LH) or fiber (LF) insulation can be ordered in a variety of sizes and maximum temperature options. Models are rated at 1200, 1300, or 1400 °C. Control options include the B500 controller (5 programs with 4 segments each), or the C540 controller (10 programs with 20 segments each).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty