Would you like to log in again?
Chamber Furnaces for Annealing, Hardening and Brazing, Nabertherm
Supplier: Nabertherm
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nabertherm
N02H1NMNA-208 N06H1NKNE-480 N02H1NKNA-208 N04H1NKNE-480 N01H1NMNA-240 N05H1NKNE-480 N05H1NMNE-480 N03R1NKNE-480 N02R1NMNE-480 N03R1NMNE-480 N06H1NMNE-480 N01H1NMNA-208 N04H1NMNE-480 N01H1NKNA-208 N02H1NKNA-240 N02H1NMNA-240 N07H1NMNE-480 N02R1NKNE-480 N07H1NKNE-480
76538-208EA 7317.15 USD InStock
76538-208 76538-218 76538-206 76538-228 76538-216 76538-226 76538-204 76538-214 76538-224 76538-202 76538-212 76538-234 76538-200 76538-222 76538-210 76538-232 76538-220 76538-198 76538-230
Chamber Furnaces for Annealing, Hardening and Brazing, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
The N 7/H - N 87/H chamber furnaces are designed to withstand harsh use in the laboratory.
  • Three-sided heating from both sides and the bottom, heating elements on support tubes ensure free heat radiation and a long service life
  • Bottom heating protected by heat-resistant SiC plate, low energy consumption due to multi-layer insulation
  • Exhaust opening in the side of the furnace, or on back wall of chamber furnace for model N 31/H and higher
  • Base frame included in the delivery, N 7/H - N 17/HR designed as benchtop model
  • Parallel guided downward swinging door (user protected from heat radiation)
  • Gas spring dampers provide for easy door opening and closing
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
The furnaces can be extended with a variety of accessories, like annealing boxes for operation under protective gas, roller guides, or a cooling station with a quench tank. Even high-performance applications such as the annealing titanium in medical applications can be implemented without the use of expensive and complicated annealing systems.
The chamber furnaces are ideal when heat-treating metals and robust insulation with light refractory bricks is necessary.
Accessories information: The wide selection of chamber furnaces for annealing, hardening and brazing can be extended with a variety of accessories to suit your application. The accessories detailed here represent only a small fraction of the products available. For further details, please see our separate catalogues for heat-treatment furnaces and hardening accessories. Optional accessories include annealing boxes; annealing tray with holder; hearth plates; hardening tongs; heat treating foil; gloves. Contact your Avantor representative for more information.
Ordering information: These chamber furnaces can be ordered with a variety of sizes and voltage options. Models are rated at 1280 °C. Control options include the B500 controller (5 programs with 4 segments each), or the C540 controller (10 programs with 20 segments each).
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty