Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High-Temperature Furnaces with MoSi₂ Heating Elements, Up to 1800 °C, LHT Model, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nabertherm
LHT226VN-240 LHT227VN-208 LHT127WN1-208 LHT226VN-208 LHT427VN-480 LHT127WN1-240 LHT228VN-208 LHT426VN-480 LHT428VN-480 LHT327VN1-208 LHT827VN-480 LHT828VN-480 LHT826VN-480 LHT227VN-240 LHT327VN1-240 LHT228VN-240
76538-258EA 18053.02 USD InStock
76538-258 76538-278 76538-288 76538-276 76538-286 76538-264 76538-274 76538-262 76538-284 76538-272 76538-260 76538-282 76538-270 76538-292 76538-280 76538-290
High-Temperature Furnaces with MoSi₂ Heating Elements, Up to 1800 °C, LHT Model, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
Designed as tabletop models, these compact high-temperature furnaces have a variety of advantages.

  • Tmax 1600, 1750, or 1800 °C
  • High-quality molybdenum disilicide heating elements
  • Furnace chamber lined with first-class, long-life fiber material, unclassified
  • Housing made of sheets of textured stainless steel
  • Dual shell housing with additional fan cooling for low surface temperature
  • Compact design with lift door, opening upwards
  • Adjustable air inlet
  • Exhaust air opening in the roof
  • Type B thermocouple
  • Switchgear with steady control of the heating elements
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive

The first-class workmanship using high-quality materials, combined with ease of operation, make these furnaces all-rounders in research and the laboratory. These high-temperature furnaces are also perfectly suited for the sintering of technical ceramics, such as zirconium oxide dental bridges.

Accessories information: Optional accessories include over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; square saggar for charging of up to three layers; protective gas connection to purge with non-flammable protective or reaction gases; manual or automatic gas supply system; process control and documentation via VCD software package for monitoring, documentation and control. Contact your Avantor representative for more information.

Ordering information: These high-temperature furnaces with MoSi2 heating elements can be ordered with a variety of sizes, temperature, and voltage options. Models are rated up to 1600, 1650, 1750 or 1800 °C. Furnaces come with the P570 controller (50 programs with 40 segments each).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty