Would you like to log in again?
Melting Furnaces, Nabertherm
Supplier: Nabertherm
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nabertherm
KD0230IRNA-240 KD0230IRNA-208 KD0430IRNA-480 KD0130IRNA-240 KC0143IENC-480 KD0110IRNA-208 KD0210IRNA-208 KD0410IRNA-240 KC0243IENC-480 KD0130IRNA-208 KD0110IRNA-240 KD0410IRNA-208 KD0210IRNA-240
76538-308EA 13698 USD InStock
76538-308 76538-318 76538-306 76538-316 76538-304 76538-314 76538-302 76538-312 76538-300 76538-310 76538-298 76538-296 76538-294
Melting Furnaces, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
These compact melting furnaces for the melting of non-ferrous metals and alloys are one of a kind and have a number of technical advantages
  • Tmax 1000, 1300 or 1500 °C, with melt temperature about 80 to 110 °C lower
  • Crucible sizes of 0.75, 1.5 or 3 liters
  • Crucible with integrated pouring spout of iso-graphite included with delivery
  • Additional spout (not for KC), mounted at the furnace for exact pouring
  • Compact bench-top design, simple emptying of crucible by tiltiing system with gas damper
  • Crucible for heating up of melting furnace insulated with a hinged lid, lid opened when pouring
Designed as tabletop models, they can be used for many laboratory applications. The practical counter balanced hinge with shock absorbers and the spout (not for KC) on the front of the furnace make exact dosing easy when pouring the melt. The melting furnaces are available for furnace chamber temperatures of 1000, 1300 or 1500 °C. This corresponds to melt temperatures of about 80 to 110 °C lower.
Accessories information: Optional accessories include other crucible types available, e.g. steel; design as bale-out furnace without tilting device, e.g. for lead melting; over-temperature limiter for the furnace chamber with automatic reset to protect against overtemperature. The limit controller switches off the heating when the pre-set limit temperature has been reached and does not switch it on again until the temperature falls below the setting again; observation hole for melt. Contact your Avantor representative for more information.
Ordering information: These melting furnaces can be ordered in a variety of temperatures, crucible sizes and voltages. Models rated up to 1000, 1300 or 1500 °C. Control through basic R7 controller with one set temperature.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty