Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Compact Tube Furnaces, Model R, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

R-472L2AN-480 R-322W3AN3-480 R-472W3AN3-480 R-472W2AN3-480 R-572L2AN-480 R-322O2AN-480 R-152L2AN-240 R-152O2AN-208 R-322O3AN-480 R-472L3AN-480 R-152L3AN-240 R-572O3AN-480 R-252L3AN-240 R-252W2AN3-208 R-252O3AN-240 R-572W2AN3-480 R-322W2AN3-480 R-252W3AN3-208 R-252W3AN3-240 R-322L3AN-480 R-252O2AN-240 R-322L2AN-480 R-572W3AN3-480 R-252O2AN-208 R-152L3AN-208 R-252L2AN-208 R-252L2AN-240 R-572L3AN-480 R-152O2AN-240 R-572O2AN-480 R-152L2AN-208 R-152O3AN-240 R-472O3AN-480 R-252L3AN-208 R-472O2AN-480 R-152O3AN-208 R-252W2AN3-240 R-252O3AN-208
76538-408EA 8740.58 USD InStock
76538-408 76538-406 76538-404 76538-348 76538-402 76538-368 76538-346 76538-400 76538-344 76538-366 76538-388 76538-352 76538-374 76538-396 76538-372 76538-350 76538-394 76538-370 76538-392 76538-390 76538-418 76538-416 76538-414 76538-358 76538-412 76538-378 76538-356 76538-410 76538-354 76538-398 76538-376 76538-364 76538-386 76538-384 76538-362 76538-382 76538-360 76538-380
Compact Tube Furnaces, Model R, Nabertherm
Ovens Tube Furnaces
These compact tube furnaces with integrated control systems can be used universally for many processes.

  • Single-zoned design as standard; Three-zoned design extends the constant temperature accuracy within the heated length
  • Dual shell housing made of sheets of textured stainless steel
  • Outer tube diameter of 50 to 170 mm, heated length from 250 to 1000 mm
  • Tmax 1200 °C: Type N thermocouple, Tmax 1300 °C: Type S thermocouple
  • Solid state relays provide for low noise operation
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive

Equipped with a standard working tube of C530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.

Accessories information: Optional accessories include: Over-temperature limiter with adjustable cutout temperature for thermal protection class 2 in accordance with EN 60519-2 as temperature limiter to protect the furnace and load; charge control with temperature measurement in the working tube and in the furnace chamber outside the tube; Three-zoned design (heated length from 500 mm); variety of working tubes and materials; gas supply systems for protective gas or vacuum operation; process control and documentation via VCD software package for monitoring, documentation and control. Contact your Avantor representative for more information.

Ordering information: These tube furnaces can be ordered in a variety of voltages, temperatures, heated lengths, Single or Three-zone control, and diameters depending on the required charge size. Select Single-zoned or Three-zoned models depending on required temperature uniformity across the needed heated length. In general, Three-zoned models extend the accurate constant temperature in the heated length from one-third of the heated length to one-half. Models are rated up to 1200 or 1300 °C. Control options include the B510 controller (5 programs with 4 segments each), C550 controller (10 programs with 20 segments each), or P580 controller (50 programs with 40 segments each).

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty