Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High-Temperature Furnaces with MoSi₂ Heating Elements, Fiber Insulation up to 1800 °C, Model HT, Nabertherm

Supplier: Nabertherm

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nabertherm
HT0357VNA-480 HT0246VNA-480 HT0248VNA-480 HC0356VNA-480 LHT827VN-480 HT0358VNA-480 HT0356VNA-480
76538-430EA 52962.72 USD InStock
76538-430 76538-428 76538-426 76538-424 76538-434 76538-422 76538-432
High-Temperature Furnaces with MoSi₂ Heating Elements, Fiber Insulation up to 1800 °C, Model HT, Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
Due to their solid construction and compact stand-alone design, these high-temperature furnaces are perfect for processes in the laboratory where the highest precision is needed.

  • Tmax 1600, 1750, or 1800 °C
  • Recommended working temperature 1750 °C (for models HT ../18), increased wear and tear must be expected in case of working at higher temperatures
  • Dual shell housing with fan cooling for low shell temperatures
  • Heating from both sides via molybdenum disilicide heating elements
  • High-quality fiber insulation backed by special insulation
  • Side insulation constructed with tongue and groove blocks provides for low heat loss to the outside
  • Long-life roof insulation with special suspension
  • Chain-guided parallel swivel door for defined opening and closing of the door
  • Two-door design (front/back) for high-temperature furnaces > HT 276/..
  • Labyrinth sealing ensures the least possible temperature loss in the door area
  • Reinforced floor as protection for bottom insulation as standard from models HT 16/16 upwards
  • Vapor vent in the furnace roof
  • Heating elements switched via thyristors
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive

Outstanding temperature uniformity and practical details set unbeatable quality benchmarks. For configuration for your processes, these furnaces can be extended with extras from our extensive option list.

Accessories information: Optional accessories include: uncontrolled or controlled cooling system with frequency-controlled cooling fan and motor-driven exhaust air flap; furnace in DB design featuring fresh air preheating, exhaust gas ventilation and an extensive safety package for debinding and sintering in one process, i.e. without transfering the material from the debinding furnace to the sintering furnace; stainless steel exhaust gas top hats; special heating elements for zirconia sintering provide for longer service life with respect to chemical interaction between charge and heating elements; protective gas connection to purge with non-flammable protective or reaction gases; manual or automatic gas supply system; inner process box to improve the gas tightness and to protect the furnace chamber against contamination; lift door; motorized exhaust air flap, switchable via the program; thermal or catalytic exhaust cleaning systems; process control and documentation via VCD software package or Nabertherm Control Center (NCC) for monitoring, documentation and control. Contact your Avantor representative for more information.

Ordering information: These high temperature furnaces with molybdenum disilicide heating elements with fiber insulation up to 1800 °C can be ordered in a variety of sizes and maximum temperature options. Models are rated up to 1600, 1750 or 1800 °C. For use with 480 V 3-phase power. Standard units come with the P570 controller that includes 50 programs with 40 segments each. Please contact your Avantor representative for more information about the various optional accessories available.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty