Would you like to log in again?
NEBNext Ultra II FS DNA PCR-free Library Prep Kit for Illumina, New England Biolabs
Supplier: New England Biolabs
NEBNext® Ultra™
E7430L E7430S E7430L E7430S
76538-650EA 2349 USD InStock
76538-650 CA76538-648 76538-648 CA76538-650
NEBNext Ultra II FS DNA PCR-free Library Prep Kit for Illumina, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEBNext Ultra II DNA PCR-free Library Prep Kit contains the enzymes and buffers required to convert a broad range of intact DNA input amounts into libraries for next-generation sequencing on the Illumina platform, all without an amplification step.
  • Includes NEBNext FS (Fragmentation System) enzymatic fragmentation reagents
  • Save time: optimized PCR-free workflow, reduced hands-on time and automation compatibility
  • Generate high quality libraries without an amplification step with inputs from 50 to 500 ng
  • Improved library uniformity
  • For use with the NEBNext Unique Dual Index UMI Adaptors DNA Set 1
The fast, user-friendly workflow includes enzymatic fragmentation for optimal input size, requires minimal hands-on time, and maintains excellent library yields and GC coverage without requiring PCR. Don’t be biased! The NEBNext® Ultra™ II FS DNA PCR-free Library Prep Kit for Illumina® can save you time and minimize amplification-induced sequence bias and errors with a complete solution for PCR-free NGS library preparation. Built on the robust and reliable Ultra II reagents and workflow from New England Biolabs®, you can trust that your samples are in the best hands – your own.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty