Would you like to log in again?
CryoSure® Vials and Caps, Caplugs
Supplier: Caplugs
222-3902-085 222-3901-085 222-3926-085 222-3905-085
76538-736CT 166.34 USD InStock
76538-736 76538-734 76538-732 76538-730
CryoSure® Vials and Caps, Caplugs
Vials
Caplugs Evergreen CryoSure® brand vials are recommended for storing cell cultures, blood/serum specimens, sperm, and other biological specimens at vapor phase* liquid nitrogen temperatures (–320 °F/–195 °C).
  • CryoSure® vials feature external threads designed to be less susceptible to aerosol contamination than internally threaded vials
  • Straight vertical inside walls with no shoulder for a large accessible opening which makes processing and retrieval of samples convenient
  • Ultra-clear polypropylene material - provides superior tensile strength and resistance to gamma radiation sterilization
  • Available in multiple colors
Caplugs Evergreen CryoSure® vial caps fit all sizes of CryoSure® vials. They have a molded-in sealing ring for rim-seal that provides a leakproof closure without the use of a silicone gasket.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty