Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® GeneMate™ Digital Magnetic Stirrer

Supplier: Avantor

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

GeneMate
H3770-S-GM
76538-826EA 347.5 USD InStock
76538-826
VWR® GeneMate™ Digital Magnetic Stirrer
Stirrers Magnetic Stirrers
GeneMate™ Magnetic Stirrer, 7×7", with stirring speed up to 1500 rpm.

  • Excellent chemical resistance with white ceramic work surface
  • Stir volumes up to 5 L
  • Safety LED warns user when top plate is unsafe to touch

Control type: Digital

Certifications: CE marked

Ordering information: Corresponding magnetic hotplate stirrers (heating and stirring 76538-822 and 76538-828) or hotplates (heating only 76538-824) with ceramic plates are also available. Full specification details are given under separate entries, please view full details by searching for the relevant product code.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty