Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Multi-position Magnetic Hotplate Stirrers

Supplier: VWR
VWR®
IPS7000-10-VWR IPS7000-5-VWR
76538-836EA 2144.72 USD InStock
76538-836 76538-834
VWR® Multi-position Magnetic Hotplate Stirrers
Stirrers Magnetic Hotplate Stirrers, Multi-position
VWR® multi-position magnetic hotplate stirrers for simulataneous heating and mixing of up to 5 or 10 vessels.

  • Simultaneous heating and stirring
  • White ceramic top surface
  • Digital control of both speed and temperature

Control : 35...120 °C
Recommended stir bar : 25 - 30 mm
Position Ø : 100 mm

Certifications: CE marked

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty