Would you like to log in again?
ABS® Undercounter and Countertop Freezers Certified to the NSF/ANSI 456 Standard for Vaccine Storage, Horizon Scientific
Supplier: Horizon Scientific
PH-ABT-NSF-UCBI-0420-LH PH-ABT-NSF-UCBI-0420 PH-ABT-NSF-UCBI-0420SS PH-ABT-NSF-UCBI-0420-ADA PH-ABT-NSF-UCFS-0120-LH PH-ABT-NSF-UCBI-0420SS-LH PH-ABT-NSF-UCFS-0120 PH-ABT-NSF-UCBI-0420-ADA-LH
76538-892EA 2330.1 USD InStock
76538-892 76538-890 76538-904 76538-902 76538-900 76538-898 76538-896 76538-894
ABS® Undercounter and Countertop Freezers Certified to the NSF/ANSI 456 Standard for Vaccine Storage, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
These laboratory and pharmaceutical freezers offer premium protection for vaccine storage.
  • Microprocessor controller with LED display
  • Integrated audible and visual alarms
  • Hermetic, high performance compressor
  • White powder coated steel construction
  • Natural hydrocarbon refrigerants
  • NSF/ANSI 456 Vaccine Storage Certification
  • ENERGY STAR certified*
These premier freezers are designed in accordance with the NSF/ANSI 456 Standard for Vaccine Storage. Units protect pharmaceuticals at optimal temperatures, preventing waste and allowing for peak delivery. The freezers utilize microprocessor controllers and feature temperature alarms, remote alarm contacts and probe access ports with included probes. American Biotech Supply Vaccine Storage Freezers utilize HFC-free refrigerant for environmental health and energy efficiency.
Certifications: Compliant with the thermal performance requirements as defined in the NSF/ANSI 456 Standard for Vaccine Storage for all testing protocols. UL, C-UL, ETL, C-ETL listed (either single or dual agency listings) and certified to UL471 standard, hydrocarbon refrigerant safety.
*Select models ENERGY STAR certified: 76538-890, 76538-892, 76538-894, 76538-896, 76538-898, 76538-900, 76538-902, and 76538-904
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty