Would you like to log in again?
AccuThrive Agility Microtome Blades with Microcoat, Stainless Steel, AccuTec Blades
Supplier: AccuTec Blades
AVBL-2006-0000 AVBL-2023-0000 AVBL-2021-0000 AVBL-2004-0000 AVBL-2007-0000 AVBL-2005-0000 AVBL-2022-0000 AVBL-2020-0000
76539-000BX 126.41 USD InStock
76539-000 76539-006 76538-998 76539-008 76539-002 76539-034 76539-004 76539-036
AccuThrive Agility Microtome Blades with Microcoat, Stainless Steel, AccuTec Blades
Microtomes and Microtome Assemblies Microtome Blades and Handles
Choose between high-profile or low-profile microtome blades for use on soft tissue.
  • Coated blades ideal for use on soft tissue
  • Also suitable for use on frozen tissue
  • Available in high-profile or low-profile formats
AccuThrive's MicroCoat blade enrichment technology adds a layer of protection for each blade, helping maintain a sharp effective edge over multiple uses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty