Would you like to log in again?
Rapid Response™ Fentanyl Test Strip Kit (Liquid / Powder), BTNX
Supplier: BTNX
FYL-18S7-5
76539-042CT 2041.99 USD InStock
76539-042
Rapid Response™ Fentanyl Test Strip Kit (Liquid / Powder), BTNX
Drug Tests
The Rapid Response™ Fentanyl (FYL) forensic test kit is a lateral flow chromatographic immunoassay for the qualitative detection of fentanyl in liquid and powder substances at the cut-off concentration of 200 ng/ml.
  • Sample: Liquid/Powder
  • Detects fentanyl at a cut-off concentration of 200 ng/mL or higher.
  • Full kit with all materials required to test drug samples
  • Results interpretation guide printed on each pouch
  • Easy to interpret results with built-in procedural controls
  • Individually packaged strips
  • Sensitive to fentanyl and analogues: Carfentanil, Butyryl Fentanyl, p-Fluoro Fentanyl, Acetyl Fentanyl, Fentanyl, Furanyl Fentanyl, Valeryl Fentanyl, Ocfentanil, 3-Methyl Fentanyl, Remifentanil, Sufentanil
Warning: This test does not evaluate drug safety or purity.
The Rapid Response™ Fentanyl Test Strip Kit (Liquid / Powder) detects the presence of fentanyl in liquid and powder substances at a cut-off of 200 ng/ml or higher.
It is specific for fentanyl screening with no significant cross-reactivity to other opiates, such as morphine and heroin. The kit includes all materials needed to perform the test: 5 individually packaged test strips, 5 plastic test tubes with extraction buffer solution, 5 swabs, a test tube holder for a mess free workspace, a result interpretation card, and detailed instructions.
To perform the test, place the desired number of buffer tubes in the tube stand. Remove the buffer tube seal and remove the swab from its packaging. Collect liquid or solid samples: dip the swab into the liquid sample or moisten the swab in the buffer tube and then wipe the swab in the substance to absorb the solid sample. Insert the swab into the buffer tube, stirring for 1-2 minutes, and then remove the swab. Remove the test strip from the pouch and hold the strip by the end where the product name is printed. Dip the strip in the liquid for 10-15 seconds. Remove the strip and place it on a non-absorbent flat surface.
A negative result can be interpreted as soon as both the test (T) and control (C) lines appear. A positive result can be interpreted once the control line has appeared and the background has cleared to white, typically by 60 seconds. Do not read results after 10 minutes. Please read the Product Insert supplied with the kit before use.
Caution: The test device is not intended to determine the purity, composition, or if the substance being examined is safe to use.
A positive or negative test result is NOT an indication that the substance being examined is safe to use. Many factors come into play when examining the samples, including but not limited to mixture of multiple substances, solubility, and pH of the sample.
BTNX Inc. does not encourage the use, supply, or production of illegal drugs or controlled substances in any way. The device is intended for harm reduction purposes. Follow the advice of your local harm reduction or public health agency.
There is a possibility that technical or procedural errors as well as other substances and factors may interfere with the Rapid ResponseTM Fentanyl Test Strip (Urine) and cause false results.
A positive result indicates the presence of fentanyl only and does not indicate quantity.
A negative result does not at any time rule out the presence of fentanyl, as they may be present below the minimum detection level of the test.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty