Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Entris® II Essential Precision Balances, NTEP-Certified, Sartorius
Supplier: Sartorius
Entris® II Essential
BCE623-1NUS BCE1202-1NUS BCE5201-1NUS BCE4202-1NUS BCE653-1NUS BCE8201-1NUS BCE6202-1NUS BCE5201-1NUS BCE622-1NUS BCE1202-1NUS BCE8201-1NUS BCE323-1NUS BCE653-1NUS BCE6202-1NUS BCE623-1NUS BCE3202-1NUS BCE3202-1NUS BCE4202-1NUS BCE622-1NUS BCE323-1NUS
76539-110EA 1514.6 USD InStock
76539-110 76539-120 CA76539-118 CA76539-106 76539-108 CA76539-108 76539-116 CA76539-114 CA76539-102 76539-104 CA76539-116 76539-118 CA76539-104 76539-106 CA76539-110 76539-112 CA76539-120 76539-114 CA76539-112 76539-102
Entris® II Essential Precision Balances, NTEP-Certified, Sartorius
Balances and Scales
100% NTEP-compliant/legal for trade weighing-Entris® II Essential line is the only precision balance in its class offering LED touch technology and 12 built-in applications.
  • Easy clean for operational efficiency and durability
  • Effective draft shield
  • Highest quality standards
  • Multilanguage (English, German, France, Italian, Spanish, Portuguese, Russian, Polish)
High chemical resistance ensured using parts made from hard-wearing polybutylene terephthalate (PBT), stainless steel and glass - prevents cross contamination with wipe clean design and easy-to-remove parts. Effective draft shield minimizes weighing errors caused by electrostatically charged samples using specially coated glass parts.
Automatically detects sartorius accessories (e.g. printer, second display). Real 'PC-direct feature' for easy connection to a PC to transfer weighing data directly into spreadsheets or documents such as Microsoft® Excel or Word.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty